Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srj-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028902.1 | NP_001024073.1 | 5 - 6 | 827 | 80 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Caenorhabditis elegans Probe spans exons |