Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313185.3 | NP_001300114.1 | 1 - 2 | 282 | 136 | |
NM_001313186.4 | NP_001300115.1 | 1 | 260 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |