Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srt-41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071105.5 | NP_503506.2 | 4 - 5 | 672 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |