Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srj-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313319.1 | NP_001300248.1 | 2 - 3 | 258 | 71 | |
NM_071138.2 | NP_503539.2 | 1 - 2 | 306 | 71 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |