Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380613.1 | NP_001368574.1 | 3 - 4 | 704 | 79 | |
NM_071160.9 | NP_503561.2 | 3 - 4 | 698 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |