Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lbp-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038422.6 | NP_001033511.1 | 2 - 3 | 314 | 101 | |
NM_001038423.5 | NP_001033512.1 | 1 - 2 | 264 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |