Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029097.5 | NP_001024268.1 | 7 - 8 | 845 | 106 | |
NM_001029098.5 | NP_001024269.1 | 6 - 7 | 782 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |