Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029097.5 | NP_001024268.1 | 9 - 10 | 1229 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |