Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038398.3 | NP_001033487.1 | 4 - 5 | 800 | 69 | |
NM_001316801.3 | NP_001303730.1 | 2 - 3 | 339 | 69 | ||
NR_133578.1 | 4 - 5 | 669 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |