Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316820.3 | NP_001303749.1 | 2 - 3 | 415 | 82 | |
NM_071223.8 | NP_503624.2 | 3 - 4 | 791 | 82 | ||
GenBank mRNA | AF273807.1 | - | 3 - 4 | 758 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |