Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srj-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071302.1 | NP_503703.1 | 6 - 7 | 661 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |