Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srt-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071305.2 | NP_503706.2 | 3 - 4 | 726 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |