Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: str-33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071340.1 | NP_503741.1 | 1 - 2 | 365 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |