Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srj-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316818.1 | NP_001303747.1 | 5 - 6 | 842 | 103 | |
NM_071355.2 | NP_503756.2 | 5 - 6 | 836 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |