Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ketn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307737.5 | NP_001294666.1 | 23 - 24 | 13560 | 73 | |
NM_001307738.4 | NP_001294667.1 | 22 - 23 | 13437 | 73 | ||
NM_001307739.4 | NP_001294668.1 | 25 - 26 | 13782 | 73 | ||
NM_001307740.4 | NP_001294669.1 | 24 - 25 | 13659 | 73 | ||
NM_001316807.3 | NP_001303736.1 | 10 - 11 | 9084 | 73 | ||
NM_001316808.3 | NP_001303737.1 | 7 - 8 | 8673 | 73 | ||
NM_001377590.2 | NP_001364781.1 | 14 - 15 | 11520 | 73 | ||
NM_001377591.2 | NP_001364828.1 | 16 - 17 | 11742 | 73 | ||
NM_001377592.1 | NP_001364782.1 | 15 - 16 | 11643 | 73 | ||
NM_001377593.1 | NP_001364783.1 | 15 - 16 | 11643 | 73 | ||
NM_001377594.2 | NP_001364827.1 | 17 - 18 | 11865 | 73 | ||
GenBank mRNA | AY819766.1 | - | 18 - 19 | 12430 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |