Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srj-49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071369.1 | NP_503770.1 | 3 - 4 | 655 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |