Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srj-55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071372.2 | NP_503773.2 | 4 - 5 | 833 | 98 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |