Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srw-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373011.2 | NP_001360542.1 | 1 - 2 | 585 | 129 | |
NM_071427.6 | NP_503828.3 | 1 - 2 | 585 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |