Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: str-68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318268.1 | NP_001305197.1 | 1 - 2 | 431 | 119 | |
NM_071450.1 | NP_503851.1 | 2 - 3 | 944 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |