Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srbc-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071495.2 | NP_503896.2 | 4 - 5 | 647 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |