Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srj-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322515.1 | NP_001309651.1 | 5 - 6 | 639 | 92 | |
NM_001322516.1 | NP_001309653.1 | 4 - 5 | 392 | 92 | ||
NM_001322517.1 | NP_001309652.1 | 4 - 5 | 363 | 92 | ||
NM_071562.2 | NP_503963.1 | 5 - 6 | 987 | 92 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |