Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt-64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322526.3 | NP_001309621.1 | 1 - 2 | 100 | 144 | |
NM_071577.9 | NP_503978.1 | 3 - 4 | 340 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |