Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp-34A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047606.5 | NP_001041071.1 | 6 - 7 | 1418 | 86 | |
NM_001047608.3 | NP_001041073.1 | 1 - 2 | 222 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |