Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: otpl-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322547.4 | NP_001309627.1 | 6 - 7 | 1418 | 118 | |
NM_001322548.3 | NP_001309628.1 | 5 - 6 | 1331 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |