Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sos-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373020.3 | NP_001360576.1 | 8 | 1576 | 92 | |
NM_071834.5 | NP_504235.2 | 8 - 9 | 1588 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |