Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pmt-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071847.7 | NP_504248.1 | 4 - 5 | 1133 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |