Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prx-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071867.5 | NP_504268.1 | 2 - 3 | 470 | 113 | |
GenBank mRNA | AB010968.1 | - | 2 - 3 | 434 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |