Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tig-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071870.5 | NP_504271.1 | 1 - 2 | 226 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |