Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pgl-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071877.6 | NP_504278.1 | 2 - 3 | 288 | 98 | |
GenBank mRNA | AB120729.1 | - | 2 - 3 | 270 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |