Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srr-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071946.9 | NP_504347.2 | 3 - 4 | 250 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |