Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sid-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071971.9 | NP_504372.2 | 3 - 4 | 627 | 98 | |
GenBank mRNA | AF478687.1 | - | 2 - 3 | 452 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |