Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt-51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071987.6 | NP_504388.2 | 2 - 3 | 409 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |