Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glc-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072040.7 | NP_504441.1 | 3 - 4 | 665 | 68 | |
GenBank mRNA | AJ243914.1 | - | 3 - 4 | 382 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |