Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glb-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307815.4 | NP_001294744.1 | 6 - 7 | 658 | 78 | |
NM_001356646.3 | NP_001343626.1 | 6 - 7 | 668 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |