Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129376.4 | NP_001122848.1 | 4 - 5 | 590 | 69 | |
NM_072093.7 | NP_504494.2 | 4 - 5 | 566 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |