Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mcp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028468.4 | NP_001023639.1 | 6 - 7 | 815 | 78 | |
NM_001380650.1 | NP_001367490.1 | 6 - 7 | 882 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |