Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhs-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047644.2 | NP_001041109.1 | 7 - 8 | 780 | 110 | |
NR_136375.1 | 10 - 11 | 1383 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |