Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rme-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029021.4 | NP_001024192.1 | 4 - 5 | 1003 | 84 | |
NM_001029023.3 | NP_001024194.1 | 2 - 3 | 294 | 84 | ||
NM_001029024.4 | NP_001024195.1 | 2 - 3 | 210 | 84 | ||
NM_001029025.6 | NP_001024196.1 | 2 - 3 | 337 | 84 | ||
NM_001356657.3 | NP_001343865.1 | 2 | 306 | 84 | ||
NM_001356658.3 | NP_001343866.1 | 2 | 306 | 84 | ||
NM_001356659.3 | NP_001343867.1 | 1 - 2 | 188 | 84 | ||
NM_001380664.2 | NP_001367910.1 | 3 - 4 | 948 | 84 | ||
NM_001383317.1 | 2 - 3 | 236 | 84 | |||
GenBank mRNA | AF357876.1 | - | 2 - 3 | 210 | 84 | |
AF357878.1 | - | 2 - 3 | 294 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |