Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rme-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029021.4 | NP_001024192.1 | 2 - 3 | 361 | 74 | |
NM_001380664.2 | NP_001367910.1 | 1 - 2 | 306 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |