Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrck-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313212.2 | NP_001300141.1 | 6 - 7 | 1979 | 95 | |
NM_001313213.3 | NP_001300142.1 | 2 - 3 | 1043 | 95 | ||
NM_001313214.3 | NP_001300143.1 | 2 - 3 | 1049 | 95 | ||
NM_072198.7 | NP_504599.2 | 6 - 7 | 1895 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |