Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072206.2 | NP_504607.1 | 6 - 7 | 939 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |