Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sago-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072209.4 | NP_504610.1 | 4 - 5 | 872 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |