Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083280.3 | NP_001076749.1 | 14 - 15 | 2586 | 66 | |
NM_001373040.1 | NP_001360140.1 | 14 - 15 | 2586 | 66 | ||
NM_001383319.1 | 14 - 15 | 2564 | 66 | |||
GenBank mRNA | AF338767.1 | - | 14 - 15 | 2573 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |