Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083280.3 | NP_001076749.1 | 6 - 7 | 710 | 100 | |
NM_001383319.1 | 6 - 7 | 688 | 100 | |||
GenBank mRNA | AF338767.1 | - | 6 - 7 | 697 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |