Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ubh-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072252.7 | NP_504653.2 | 6 - 7 | 1420 | 103 |
Gene Symbol: W01A11.7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072251.6 | NP_504652.2 | 3 - 4 | 550 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |