Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ubh-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072253.5 | NP_504654.1 | 3 - 4 | 263 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |