Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: K04A8.21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072264.4 | NP_504665.3 | 1 - 2 | 82 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |