Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072282.5 | NP_504683.1 | 8 - 9 | 1228 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |