Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dbl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072308.8 | NP_504709.1 | 7 - 8 | 1181 | 70 | |
GenBank mRNA | AF004395.1 | - | 7 - 8 | 1139 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |