Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpa-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072344.3 | NP_504745.1 | 2 - 3 | 183 | 86 | |
GenBank mRNA | AY008132.1 | - | 2 - 3 | 175 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |